Gas Cylinder Forklift Pallets, End Loaded, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
These End Loaded Gas Cylinder Forklift Pallets provide safety when moving and storing of gas cylinders.
- Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
- Textured finish on the pallet provides a non-skid surface
- Secure transportation of multiple gas cylinders with end-load oriented pallets
A group of gas cylinders can be secured using the 4536 kg (10000 lb.) DOT compliant strap that attaches to frame at either 305 mm, 610 mm, or 915 mm (12", 24" or 36") from the deck.
Fully welded construction from 11 gauge (3 mm) hot-rolled steel and structural tube steel, powder-coated to ensure longevity and chemical resistance.
For easier loading and unloading, pallets are also available with a 406 mm (16") hinged ramp.
Once loaded, ramp lifts up to lie flat against the unit for compact storage.
Certifications: Forklift pallets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Spécifications
- Catalog No:
- 76292-338
- 76292-336
- 76292-340
- 76292-334
- 76292-344
- 76292-342
- Cylinder type:
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Material:
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Mounting type:
- Forklift
- Forklift
- Forklift
- Forklift
- Forklift
- Forklift
- Straps:
- 1 Total
- 1 Total
- 1 Total
- 1 Total
- 1 Total
- 1 Total
- Strap type:
- Ratchet Binding
- Ratchet Binding
- Ratchet Binding
- Ratchet Binding
- Ratchet Binding
- Ratchet Binding