Powder Funnels with 60° Side Angle, Sati International
Fournisseur: CHEM SCIENCE INC CA
Certificats
A propos de cet article
Funnels feature a 60° side angle and a standard taper inner joint.
Customers Who Bought This Also Bought
Funnels feature a 60° side angle and a standard taper inner joint.