Trio.Bas™ Mini Kit, Orum International, Hardy Diagnostics
# de catalogue 76308-448
Fournisseur: Hardy Diagnostics
Spécifications
- Flow rate:100 l/min.
- Description:Trio.Bas™ Mini Kit
- Cat. no.:76308-448
- Head Type:CompactDry™
Spécifications
A propos de cet article
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
- Aspirating head with easy manipulation bayonet closure
- Long lasting battery
- Ergonomic and balanced design to facilitate handling with ease
- Antibacterial techno-polymer shock-proof body
- IP65 protected from dust and water
Microbial impact air sampler specifically designed to hold CompactDry™ plates and sample at a 100 liters/minute flow rate.
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.
Per USP 797, "Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.
Customers Who Bought This Also Bought