Sealing Film Dispenser, Diversified Biotech
Catalog # 10014-778
Supplier: Diversified Biotech
Specifications
- Color:Clear
- Description:Sealing Film Dispenser
- Cat. no.:10014-778
- W×D×H:165.1×101.6×50.8 mm
- Supplier No.:SFBX-1000
Specifications
About this item
These film dispensors are hinged, clear acrylic sealing film storage box with non-slip feet.
- Stores and dispenses microplate sealing film
- Conveniently remove films one-at-a-time with one hand
Customers Who Bought This Also Bought