Cryogenic Gas Cylinder Barricade Rack, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
This cryogenic gas cylinder barricade rack provides safety when moving and storing of gas cylinders.
- Secure cryogenic cylinders up to 83.8 cm (33") in diameter
- For indoor or outdoor cryogenic cylinder storage
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Barricade racks feature two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
Barricade racks surround cylinders with 51 mm (2") square steel structural tubing, continuously seam welded at all junction points and sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability.
Pre-drilled 14 mm (0.531") diameter floor mounting holes included on lower tube structure.
Certifications: Barricade racks are NFPA 1 and NFPA 55 approved, OSHA approved, CGA-P1 approved, and High Hazard Seismic Zone approved. Meets USGS high-hazard seismic zone requirements when properly mounted.
Customers Who Bought This Also Bought