DURAN® HPLC Bottles, GL 45, DWK LIFESCIENCES
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Complete system comprising of DURAN® pressure plus laboratory bottle with four port screw cap (PP), four screw connections (black, M8 thread) and silicone seals.
- Easy-to-read scale and large labelling field for easy marking
- Unused ports can be sealed with silicone blanking seals
- Autoclavable at 121 °C
Connection of different hose diameters (1.6 and 3.2 mm) as well as sterile pressure equalization sets (syringe filter 0.2 µm) is possible. It is safe to transfer of liquid media within a closed and sterile system (evaporation is reduced).
Certifications: USP standard.
Customers Who Bought This Also Bought