Sample Vials, Clear, Without Caps, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Screw thread sample vials are manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Class A requirements.
Ordering information: Closures are not included.
Packaging: Vials are packaged in corrugated trays with partitions.
Customers Who Bought This Also Bought