Storage Cabinet for Vials and Closures, Chemglass
# de catalogue 75820-616
Fournisseur: Chemglass
Spécifications
- Description:Storage cabinet with 9 drawers
- Cat. no.:75820-616
- Supplier No.:CV-5730-0009
Spécifications
A propos de cet article
This storage cabinet provides clean, organized vial and closure storage.
- Powder coated finish
- Features nine drawers
Drawer dimension: W17×11×H11"
Ordering information: Vials and closures must be purchased separately.
Customers Who Bought This Also Bought