A propos de cet article
Monitor RH and temperature inside an environmental chamber or clean room without opening the door. Place the weatherproof (IPX3) sensor outdoors, in green houses, warehouses, air ducts and other hard to reach places and continuously monitor ambient RH and temperature from a remote location. The wireless sensors broadcast to the monitor from up to 100 feet (30 m) away without obstructions, using a wireless RF signal.
The monitor displays the local and remote temperatures in ºC or ºF, recalls min/max on both, and indicates when the batteries are low in both the sensors and monitor. It also displays user adjustable, audible and visible, Hi/Lo Alarms, on both temperature and RH, for the base unit and the remote sensors.
Monitor dimension: 80×80×30 mm.
Sensor dimension: 110×60×30 mm.
Monitor weight: 113 g (4 oz).
Sensor weight: 113 g (4 oz) with batteries.