Shelf Bins
Fournisseur: Treston
Certificats
A propos de cet article
These shelf bins provide an ideal way of storing small/medium sized items in maintenance, production and stores.
- Particularly suitable for use in small parts retrieval systems and mobile shelving
- Bright colors including gray are available for easy color coding systems
- Available in four sizes in depths of 11.81, 15.75, 19.69 or 23.62"
Crystal clear bins, even when using the index label provided, allow visible identification of bin contents. Corrugated bases ease the picking of very small items. Full width label with protective shield is easy to keep clean and change when necessary. The label is included in the price.
Ordering information: Installation is required. Assembly videos available.
Delivery: Labels and dividers sold separately. Shelf bins sold in case quantities only. Optional accessories are width dividers (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought