ValueWare® Reusable Soda Lime Glass Petri Dishes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These shallow glass dishes are manufactured from soda lime glass.
Sold in sets consisting of top and bottom dishes.
it is autoclavable at 121 °C and can withstand temperatures up to 500 °C.
Customers Who Bought This Also Bought