Excellence Benchtop Refractometers, Mettler-Toledo
Fournisseur: Mettler Toledo
Certificats
A propos de cet article
Developed for a wide range of applications, the intuitive Excellence Refractometers are the next generation of refractive index measurement.
- Color touchscreen with 7" display
- Compatible with nine languages
- Accuracy up to five digits
- One Click user interface
- Simultaneous determination of density, refractive index, pH / conductivity and color
- Internal storage of up to 200 results
Brix, refractive index and more: METTLER TOLEDO refractometers master measurements of a wide variety of challenging samples with high repeatability and an accuracy of up to five decimal places.
Ordering information: Can be further enhanced with the following Optional modules and accessories to fit your process needs: powerful autosamplers, LabX PC software, ErgoSens infrared motion sensor. Contact your Avantor representative for more information.
Spécifications
- Catalog No:
- 76300-940
- 76300-942
- Display:
- 7" LCD display with color touchscreen
- 7" LCD display with color touchscreen