Melting Point Capillary Tubes
Fournisseur: Wards
Certificats
A propos de cet article
Borosilicate glass capillary melting tubes are 4" in length and have a 0.05" outer diameter capillary tubes that are particularly useful for determining the melting point of various substances.
The 100 capillary tubes come in a cylindrical container that has a hole in the lid for easily obtaining one tube at a time.
Customers Who Bought This Also Bought