Polypropylene Stoppers
Fournisseur: Wards
Certificats
Certificats
A propos de cet article
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Fournisseur: VWR International
240,47 $ to 8 653,27 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
359,30 $
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
919,35 $ to 933,31 $