VWR® Laboratory Installation SafetyWasteCaps
Fournisseur: Avantor
Certificats
A propos de cet article
These caps are mainly used as connection for exhaust filter and electronic level control sensor.
- Resistant against aggressive media
These caps are electrically conductive in nature.
Customers Who Bought This Also Bought