Superfrost® Cytogenetics Slide, Erie Scientific (Thermo Scientific)
# de catalogue 48404-468
Fournisseur: Erie Scientific
Spécifications
- Description:Cytogenetics slide
- L×W:25×75 mm (1×3")
- Thickness:1.0 mm
- Cat. no.:48404-468
- Supplier No.:CGN-4951-006
Spécifications
A propos de cet article
Superfrost® slide feature a glass surface specifically processed and inspected for cytogenetic applications.
Slide features include consistent, high-quality surface for reliable chromosome spreads, hydrophilic surface improves retention of metaphase cells and Superfrost® white tab for permanent marking.
Customers Who Bought This Also Bought