Superlite Folding Truck, Wesco International
# de catalogue 14231-232
Fournisseur: Wesco
Spécifications
- Width:19"
- Height:42"
- Material:Aluminum
- Cat. no.:14231-232
- Load Capacity:175 lbs.
- Depth:19"
- Platform Size:19×12"
- Weight:11.5 lbs.
- Supplier No.:220617
- Folded Size:19W×2D×29.5"H
- Wheels/Feet:7×1.5" rubber tread/poly hub (2)
Spécifications
A propos de cet article
Space saving folding design. Fits in to small storage spaces or car trunks.
- Elastic straps for securing load
- Aluminum construction
Non-marking, shock absorbing, rubber tread tires. Mini mover fits into overhead airline compartments.
Customers Who Bought This Also Bought