Ward's® Pipette Tip Disposal Box
Ward's Pipette Tip Disposal Box, 154×171×206 mm
470311-178
: HS234634W
Quick flip of the port cover makes tip ejection easy.
- Large, hinged lid allows easy disposal of contents
- 3/8" (10 mm) thick acrylic provides protection from Beta radiation
The ward's pipette tip disposal box is convenient for one-handed disposal of used pipette tips, and eliminates any need to handle pipette tips used with hazardous fluids. Biohazard disposal bags can be used in conjunction with this box if hazardous fluids are not acrylic friendly.
- Désignation:Ward's Pipette Tip Disposal Box
- Dimensions:154×171×206 mm