KIMBLE® HI-VAC® Right-Angle Valve, with PTFE Plug and without Tip O-Ring, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This high-vacuum valve is for use in vacuum applications up to 5×10-7 Torr.
- Similar to KT826510 series but without tip o-ring exposure
- Designed with a precision external thread for fine control
- Can be used as a bleed up to 1/2 turn
- Full open position is at 11/2 turns
- Primary seal is made with a FKM o-ring on a tapered seat
- Non-rotating PTFE shaft can be used continuously at system temperatures of 230 °C or intermittently at 275 °C
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought