KEMRESIN® Venturi Work Tops, Kewaunee®
Fournisseur: Avantor
Certificats
A propos de cet article
Venturi fume hood worktop can be used wherever optimum physical and chemical resisting properties are required.
- Ultra-smooth non-porous surface
- Composed of 1" and ³/₄" thick molded epoxy resin
- Resistant to all alkalis, acids and solvents
- Highly resistant to heat and physical abuse
It also contains features such as left rear cupsink, horizontal flush.
Certifications: Meet SEFA 3 chemical resistance performance standards.
Ordering information: Available in the colors: black, putty, gray, slate.
Customers Who Bought This Also Bought