Dura Heavy Duty Polyurethane Chairs
Fournisseur: BevCo Precision Manufacturing
Certificats
A propos de cet article
Heavy-duty chairs for the toughest environments.
- Black polyurethane seat and back are impervious to water, oil, and most chemicals
- Seat and back are also puncture-resistant and easy to clean
- Contoured back is fully adjustable encouraging proper spinal alignment
- Large waterfall seat aids in lower body circulation, improving overall wellness
- Pneumatic height adjustment allows you to easily change your seat height
- 350 lb.weight capacity
The Dura series is specifically designed to be the most durable polyurethane chair offering the greatest total body support. The advanced ergonomic features improve productivity and reduce fatigue for outstanding comfort and long-lasting use in the toughest work environments. Rugged, self-skinned polyurethane seating provides excellent cushioning properties while offering high-impact resistance. The generous seat size offers plenty of room for all-day comfort while the backrest provides ideal lumbar support to be highly effective throughout the day. Independent testing has confirmed the Dura Series provides the optimal ergonomic seating experience.
Certifications: ANSI/BIFMA; GreenGuard Gold.
Ordering information: 12-year warranty.
Spécifications
- Catalog No:
- 76248-894
- 76326-166
- 76326-168
- 76326-364
- 76326-170
- 76326-366
- 76326-172
- 76326-368
- Seat/back material:
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Polyurethane
- Weight capacity:
- 350 lbs.
- 350 lbs.
- 350 lbs.
- 350 lbs.
- 350 lbs.
- 350 lbs.
- 350 lbs.
- 350 lbs.