Biohazard Waste Bags, Autoclavable, Medegen Medical Products
Fournisseur: Medegen Medical Products, LLC
Certificats
A propos de cet article
These autoclave bags are built to withstand the rigors of steam sterilization while meeting the requirements for the collection, storage, transport and processing of biohazardous waste.
- Standard autoclave bags are tested to exceed 165 g of dart impact for compliance in the majority of US states
- Biohazardous labeling in both English and Spanish
- International Biohazard Symbol in easy-to-see contrasting color
Delivery: Twist ties included.
Packaging: Flat pack.
Caution: Not for use in CA, FL or PA.
Customers Who Bought This Also Bought