Class 100 Accelerator Free Nitrile Gloves, TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
Rival CR 9.5" ambidextrous Class 100 nitrile gloves are soft modulus and accelerator-free for those with skin sensitivities.
- Ambidextrous
- Powder-free
Techniglove products are 100% nitrile, contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. Rival CR's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide an excellent grip on all surfaces.
Suitable for use in cleanrooms and contamination controlled work environments, pharmaceutical, medical device, disc drive, and semiconductor manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag; 10 bags per case.
Spécifications
- Catalog No:
- 76302-806
- 76302-808
- 76302-802
- 76302-804
- 76302-810
- 76302-812
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips