Reactor Base System for Ethanol Extraction/Winterization, Ace Glass Incorporated
# de catalogue 76291-812
Fournisseur: Ace Glass
Spécifications
- Description:Jacketed Reactor Base System
- Cat. no.:76291-812
Spécifications
A propos de cet article
Delivery: Includes the jacketed glass vessel, head, clamps, stir shaft, agitators, and a support stand configured for an explosion proof motor.
Customers Who Bought This Also Bought