A propos de cet article
Excellent cleanroom, ESD and ESD/cleanroom options.
- Backrest is specifically shaped to provide ample back support
- Backrests have a heavy-duty polypropylene back shell to protect the upholstery
- Generous seat provides an exceptional seating experience
- Thick, resilient, high-density foam provides long-lasting comfort
- The pneumatic height adjustment allows you to easily change your seat height
- Sturdy polished aluminum base
- Mid and tall height models come with a 20" adjustable chrome footring
- 300 lbs. weight capacity
BEVCO’s Integra Series is an upholstered chair that offers around-the-clock reliability designed to meet the needs of a variety of workplace environments. This upholstered seating solution provides the most versatility, durability and overall seating enjoyment. The Integra makes working easy!
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76243-550
- 76252-486
- 76246-590
- 76248-982
- 76251-806
- 76243-272
- 76246-202
- 76246-200
- 76277-140
- 76240-930
- 76246-620
- 76247-450
- 76247-448
- 76247-692
- Model:
- 76243-550
- 76252-486
- 76246-590
- 76248-982
- 76251-806
- 76243-272
- 76246-202
- 76246-200
- 76277-140
- 76240-930
- 76246-620
- 76247-450
- 76247-448
- 76247-692
- Seat/back material:
- Vinyl
- Vinyl
- Fabric
- Vinyl
- Vinyl
- Fabric
- Vinyl
- Fabric
- Fabric
- Vinyl
- Vinyl
- Vinyl
- Fabric
- Vinyl
- Weight capacity:
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.
- 300 lbs.