NSH Series Couplings, Colder Products Company
Fournisseur: Colder Products
Certificats
A propos de cet article
NSH Series couplings are general purpose versions of our pressure-balanced, non-spill, ChemQuik® CQG couplings. Their non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety in a very low-cost package.
- Operating temperature: 0 to 49 °C
- Eliminates source of metallic contaminants
- Helps prevent accidental misconnects
- Ultimate protection from chemicals and fumes
Molded polypropylene, EPDM seals and a 100% springless and metal-free flow path provide broad chemical resistance and exceptionally high flow capacity, allowing instant disconnects (and reconnects), even under pressure.
Customers Who Bought This Also Bought