Ammonite Fossil Replica
# de catalogue 470306-900
Fournisseur: Wards
Spécifications
- Description:Replica Ammonite
- Cat. no.:470306-900
Spécifications
A propos de cet article
Ammonites grew their shells from calcium carbonate and grew constantly throughout their life span. A relative of octopi and squids today, ammonites swam through the open ocean, preying on smaller marine animals.
- Fossil replica of an ammonite from the Mesozoic measuring approximately 1.5" (4 cm)
- This fossil replica is made from a resin designed to last through heavy classroom and laboratory use
- Ammonites like this one swam through the ocean while dinosaurs roamed the earth
- Perfect for use in the classroom during lessons on fossils and paleontology
- Ammonite shell segments can be counted to determine the age of the animal
This fossil replica is perfect for lessons on paleontology and geology. It's also great for use in a fossil dig activity.
Customers Who Bought This Also Bought