Glas-Col® Fabric Insulating Tops, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Fabric insulating tops for 1, 2, and 3 neck flasks. Lacing closure allows installation without disturbing attached glassware or equipment. Insulating tops have no electrical connections.
Customers Who Bought This Also Bought