WHEATON® Headspace Crimp Top Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Headspace vials are manufactured from clear Type I borosilicate glass.
- Accept 20 mm aluminum seals
- Flat bottom and beveled finish
Ordering information: Caps not included (sold separately).
Customers Who Bought This Also Bought