Labels, Direct Thermal Cryo-Tags®, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Polypropylene labels accepts printing from direct thermal printers.
- Withstands temperature from −196 to +80 °C
- Freezable in liquid and vapor phase nitrogen
- Adhere to most plastics, glass and metals
Tubes labelled with these labels easily slide in and out of centrifuge rotors without binding.
These labels accepts writing from most lab markers including solvent resistant pens and teeny writers.
Caution: Prolonged exposure to alcohol may cause material discoloration.
Customers Who Bought This Also Bought