ContactUsLinkComponent
American Chemical Society: Glow It Up Lab Activity
American Chemical Society: Glow It Up Lab Activity
# de catalogue 470310-420
Fournisseur:  Ward's Science
Kit Glow It Up Lab Activity
American Chemical Society: Glow It Up Lab Activity
# de catalogue 470310-420
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Glow It Up Lab Activity
  • Cat. no.:
    470310-420
Spécifications
A propos de cet article
How does a glow stick work?
  • Chemiluminescence
  • Students alter the chemical reaction to attempt to change duration and brightness of the reaction
This lab, developed by the American Chemical Society through grants from the National Science Foundation (NSF) and National Institutes of Health (NIH), explores chemical reactions that release energy in the form of light, known as chemiluminescence. Teachers begin with a demonstration that uses a luminol mixture and oxidizer to start a chemical reaction that glows.
Students will then conduct an experiment to see how the amount of oxidizer affects the brightness and duration of the light. Students will also add a fluorescent highlighter dye to the reaction to see how this affects the brightness and duration of the glow. Additionally, they will learn about the invention of the glow stick and relate their experience to how a glow stick works.
Customers Who Bought This Also Bought