ContactUsLinkComponent
American Chemical Society: Chemistry's Colorful Clue Lab Activity
American Chemical Society: Chemistry's Colorful Clue Lab Activity
# de catalogue 470310-418
Fournisseur:  Ward's Science
Kit Chemistry Colorful Clue Lab Activity
American Chemical Society: Chemistry's Colorful Clue Lab Activity
# de catalogue 470310-418
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chemistry's Colorful Clue Lab Activity
  • Cat. no.:
    470310-418
Spécifications
A propos de cet article
Real world application of chemistry on natural water sources.
  • Chemistry and environmental science combined
  • Acid/base chemistry
  • pH scale
This lab activity is developed by the American Chemical Society through grants from the National Science Foundation (NSF) and National Institute of Health (NIH). Through the lessons in this lab, students will learn how human activity may affect natural water sources. Using an indicator, students will determine how acids and bases impact the pH of water.
Upon completion of this activity, students will then create carbon dioxide gas through a chemical reaction and determine if the carbon dioxide could have affected the pH of a local stream.
Customers Who Bought This Also Bought