Spécifications
- Color:Gray
- Description:Taped Embedding Cassettes with Lid
- Cat. no.:76269-682
- Shape:45° Angle
Spécifications
A propos de cet article
Avantik Taped biopsy cassettes with lid are made of the highest quality and provide great convenience.
- Taped cassettes are easy to load and are optimized for use with Leica and Sakura printers
- Cassettes feature a 45° angled area for optimal printing
- Routine, Procession, Biopsy, and Micromesh with lids fixed open, hinged, or separate
- Efficient slots ensure excellent fluid exchange and drainage
With Avantik’s EZ-I.D. label color-coding packaging system, it's easy to find the right color and style cassette. Time and care are taken to color code the individual taped cassette boxes and cases, so the contents are easily identifiable from the outside. A great convenience to laboratories carrying multiple styles of cassettes. Along with the great benefit of the color-coded system, Avantik Taped Cassettes are designed to provide the best results.