KoolTemp® GTS PUR Shippers, Cold Chain Technologies
Fournisseur: Cold Chain Technologies
Certificats
A propos de cet article
Universal, prequalified systems for domestic and international shipments of valuable vaccines, pharmaceutical products, and blood.
- 48 hour PUR shipping system for high value payloads
- Easy pack-outs using Koolit® bricks
- Pre-qualified for minimum and maximum loads
- Environmentally safe-reusable shipper contains no ozone-depleting chemicals
Pre-qualified for time/temperatures: 2 to 8 °C for 52 hours under hot conditions and 2 to 8 °C for 72 hours under cold conditions.
Customers Who Bought This Also Bought