Order Entry
ContactUsLinkComponent
Anti-HER2 Recombinant Monoclonal Antibody
Anti-HER2 Recombinant Monoclonal Antibody
# de catalogue 76262-634
Recombinant Anti-HER2 Fab Monoclonal Antibody 50UG
Anti-HER2 Recombinant Monoclonal Antibody
# de catalogue 76262-634
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HER2
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at 2-8 °C. Dilute only prior to immediate use.
  • Shipping temperature:
    Wet Ice
  • Purification:
    >90%
  • Cat. no.:
    76262-634

Spécifications

A propos de cet article

HER2 (human epidermal growth factor receptor 2) is a member of the epidermal growth factor receptor (HER/EGFR/ERBB) family. It is also called ERBB2, CD340 or proto-oncogene Neu. The protein is a receptor tyrosine kinase located on the plasma membrane of cells. Activation of the tyrosine kinase promotes cell proliferation and suppresses apoptosis. HER2 can dimerize with any other member of the ErbB family, which results in auto phosphorylation of tyrosine residues and activation of signal pathways. HER2 is over expressed in a significant proportion of cancer patients, resulting in an increased disease recurrence and poor prognosis. Therefore an early diagnosis of the HER2 status of the tumors is necessary to decide the appropriate treatment. Besides in breast cancer HER2 is over expressed in variety of different tumors. The over expression of HER2 makes it a target for immunotherapy of certain tumor patients. The antibody Trastuzumab (Herceptin) which recognizes HER2 was approved for the treatment of HER2 positive breast cancer patients in 1998. The original approval was for treatment of HER2 positive metastatic breast tumors. Later studies have shown that Herceptin is also beneficial for early stage HER2 positive tumor patients and it is approved as an adjuvant treatment. Additionally the antibody is now approved also for other HER2 positive tumors (metastatic gastric adenocarcinoma). This antibody is made from a humanized Fab fragment making it specific, efficient, and effective. Humanized Recombinant Anti-HER2 Fab fragment Antibody is useful for researchers interested in Cancer research.

Humanized Recombinant Anti-HER2 Fab fragment Antibody has been tested for use in Flow Cytometry, Western Blot, and ELISA. This antibody recognizes structured HER2 and will work in western blot when the protein has not been denatured with DTT or bMe. Although not tested, this antibody could be useful in in IHC and in in-vivo and other cellular assays. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HER2
Clonality: Recombinant Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Customers Who Bought This Also Bought