Express Transfer Buffer
# de catalogue 76236-400
Fournisseur: Rockland Immunochemical
Numéro CAS:
Spécifications
- Description:Express Transfer Buffer
- Size:1 L
- Cat. no.:76236-400
Spécifications
A propos de cet article
Express Transfer Buffer
- Western blot transfer solution
- Alternative to tris-glycine
- Lower background
Western Blotting is the method used for immunodetection for the detection of proteins. Blotting is a method for transferring biological molecules (e.g., proteins, nucleic acid fragments) from a gel matrix to a membrane support for the subsequent detection of those molecules.
Application Note: Express Transfer Buffer is a alternative to tris-glycine as a transfer buffer in the western blot. It may run at an elevated temperature but gives markedly lower background.
Customers Who Bought This Also Bought