6 Position TrueBlot® Magnetic Separator, Rockland Immunochemicals
Fournisseur: Rockland Immunochemical
Certificats
A propos de cet article
6x1.5mL Position Magnetic Separator
- magnetic separation
- six 1.5 mL microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1.5 mL microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 mL.
Magnetic separator unit can be used to hold, and separate magnetic particles.
Spécifications
- Catalog No:
- 76237-656
- 76237-654
- 76237-652
- Shipping conditions:
- At ambient temperatures
- At ambient temperatures
- At ambient temperatures