A propos de cet article
Horse Red Blood Cells 10% Washed Pooled Cells
- aseptically collected and prepared to order
- Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS)
Horse whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Horse red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Horse red blood cells are perishable and are collected and processed upon receipt of your order.
Complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- RLR409-0050
- RLR409-0100
- Shipping temperature:
- Wet Ice
- Wet Ice