Order Entry
ContactUsLinkComponent
Mouse Hemojuvelin ELISA Kit, Rockland Immunochemicals
Mouse Hemojuvelin ELISA Kit, Rockland Immunochemicals
# de catalogue 76236-216
Numéro CAS:  
Mouse Hemojuvelin ELISA Kit, Rockland Immunochemicals
# de catalogue 76236-216
Numéro CAS:  

Spécifications

  • Assay Type:
    Sandwich
  • Reactivity:
    Mouse
  • Description:
    Mouse Hemojuvelin ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-216

Spécifications

A propos de cet article

Mouse Hemojuvelin AccuSignal ELISA Kit

  • Natural and recombinant mouse Hemojuvelin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q33-D393

Hemojuvelin (HJV/RGMc/HFE2) is a membrane-bound and soluble protein in mammals. It is a member of a three gene family (in vertebrates) called the repulsive guidance molecules. In mouse, the Hemojuvelin protein is encoded by the HFE2 gene, and it is mapped to Chromosome 3. It was concluded that downregulation of hepatic Hemojuvelin during inflammation may induce a temporary elimination of iron sensing. Hemojuvelin is a coreceptor for bone morphogenetic proteins (BMPs), and inhibition of endogenous BMP signaling reduces hepcidin expression and increases serum iron in mice. Furthermore, Hemojuvelin may play inhibitory roles in prostate cancer by suppressing cell growth, adhesion, migration and invasion.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Hemojuvelin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Hemojuvelin standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought