Order Entry
ContactUsLinkComponent
Human MIA ELISA Kit, Rockland Immunochemicals
Human MIA ELISA Kit, Rockland Immunochemicals
# de catalogue 76235-692
Numéro CAS:  
Human MIA ELISA Kit, Rockland Immunochemicals
# de catalogue 76235-692
Numéro CAS:  

Spécifications

  • Reactivity:
    Human
  • Description:
    Human MIA ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-692

Spécifications

A propos de cet article

Human MIA AccuSignal ELISA Kit

  • Natural and recombinant human MIA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G25-131Q

Melanoma Inhibiting Activity(MIA), also known as cartilage-derived retinoic acid-sensitive protein(CD-RAP), is an approximately 11-15 kDa protein that is expressed as a noncovalent homodimer. MIA is structurally related to OTOR/Otoraplin and MIA-2 in a small family of secreted proteins with one SH3 domain(1-3). And the MIA gene was mapped to 19q13.32-q13.33 by fluorescence in situ hybridization. Beside, It is a marker for malignant melanoma. MIA functions as a chemoattractant for mesenchymal stem cells and enhances their BMP-2 and TGF-beta3 induced differentiation into chondrocytes . MIA-deficient mice exhibit delayed chondrocyte differentiation but enhanced chondrocyte proliferation and cartilage repair .

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human MIA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or saliva to each empty well. It is recommended that each human MIA standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought