Spécifications
- Assay Type:Sandwich
- Reactivity:Human
- Target protein:BMP-7
- Description:Human BMP-7 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-662
Spécifications
A propos de cet article
Human BMP-7 AccuSignal ELISA Kit
- Natural and recombinant human BMP-7.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S293-H431
Bone morphogenetic protein 7 or BMP7(also known as osteogenic protein-1 or OP-1) is a protein that in humans is encoded by the BMP7 gene.The protein encoded by this gene is a member of the TGF-beta superfamily. Like other members of the bone morphogenetic protein family of proteins, it plays a key role in the transformation of mesenchymal cells into bone and cartilage. It is inhibited by noggin and a similar protein, chordin, which are expressed in the Spemann-Mangold Organizer. BMP7 may be involved in bone homeostasis. It is expressed in the brain, kidneys and bladder.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human BMP-7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human bone tissue, cell culture supernates, serum, plasma((heparin, EDTA) or urine to each empty well. It is recommended that each human BMP-7 standard solution and each sample be measured in duplicate.