Spécifications
- Reactivity:Human
- Description:Human DAN - NBL1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-302
Spécifications
A propos de cet article
Human DAN - NBL1 AccuSignal ELISA Kit
- Natural and recombinant human DAN.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A17-D181
Differential screening-selected gene aberrative in neuroblastoma (DAN) is a member of the DAN family of secreted glycoproteins that are putative BMP antagonists. The NBL1 gene, also known as DAN, is originally cloned from a normal rat fibroblast cDNA library by a differential screening method. The human DAN gene is mapped to chromosome 1p36.13-p36. It is found that the DAN gene possesses a tumor suppressive activity when overexpressed in v-src transformed cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human DAN standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human DAN standard solution and each sample be measured in duplicate.