ContactUsLinkComponent
Human BMP-4 ELISA Kit, Rockland Immunochemicals
Human BMP-4 ELISA Kit, Rockland Immunochemicals
# de catalogue 76237-118
Numéro CAS:  
Human BMP-4 ELISA Kit, Rockland Immunochemicals
# de catalogue 76237-118
Numéro CAS:  
Spécifications
  • Assay Type:
    Sandwich
  • Reactivity:
    Human
  • Target protein:
    BMP-4
  • Description:
    Human BMP-4 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-118
Spécifications
A propos de cet article
Human BMP-4 AccuSignal ELISA Kit
  • Natural and recombinant human BMP-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S293-R408
Bone morphogenetic protein 4 is a protein that in humans is encoded by the BMP4 gene which is located to 14q22-q23.1, The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. BMP4 is a polypeptide belonging to the TGF-beta superfamily of proteins. It, like other bone morphogenetic proteins, is involved in bone and cartilage development, specifically tooth and limb development and fracture repair. It has been shown to be involved in muscle development, bone mineralization, and uteric bud development. BMP4 has also been implicated in Fibrodysplasia Ossificans Progressiva in which it is underexpressed. In human embryonic development, BMP4 is a critical signaling molecule required for the early differentiation of the embryo and establishing of a dorsal-ventral axis. BMP4 is secreted from the dorsal portion of the notochord, and it acts in concert with sonic hedgehog(released from the ventral portion of the notochord) to establish a dorsal-ventral axis for the differentiation of later structures. BMP4 stimulates differentiation of overlying ectodermal tissue. Inhibition of the BMP4 signal(by chordin, noggin, or follistatin) causes the ectoderm to differentiate into the neural plate. The standard product used in this kit is recombinant BMP-4 with the molecular mass of 26KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human BMP-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human bone tissue or cell culture supernates to each empty well. It is recommended that each human BMP-4 standard solution and each sample be measured in duplicate.
Customers Who Bought This Also Bought