Biotin Anti-Goat IgG Antibody for MaxTag™ Histo for IHC Kit, Rockland Immunochemicals
# de catalogue 76235-540
Fournisseur: Rockland Immunochemical
Numéro CAS:
Spécifications
- Conjugation:Biotin
- Description:Biotin anti-Goat IgG Antibody MaxTag™ Histo for IHC
- Size:1 Each
- Shipping temperature:Wet Ice
- Cat. no.:76235-540
Spécifications
A propos de cet article
Anti-Goat IgG Antibody Biotin Conjugated for IHC
- Prepared from normal serum by a multi-stage process which includes delipidation, salt fractionation and ion exchange chromatography
- Specifically designed for the use in Rockland MaxTag™ Histo for Immunohistochemistry Kit
Biotin anti-Goat IgG Antibody reagent is specifically designed for the use in Rockland MaxTag™ Histo for Immunohistochemistry Kit.
Customers Who Bought This Also Bought