ContactUsLinkComponent
Chemiluminescent Western Blot Kit: DYKDDDDK (FLAG®) Chemiluminescent Western Blotting Kit, Rockland Immunochemicals
Chemiluminescent Western Blot Kit: DYKDDDDK (FLAG®) Chemiluminescent Western Blotting Kit, Rockland Immunochemicals
# de catalogue RLKCA383
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Chemiluminescent Western Blot Kit: DYKDDDDK (FLAG®) Chemiluminescent Western Blotting Kit, Rockland Immunochemicals
# de catalogue RLKCA383
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chemiluminescent Western Blot Kit: DYKDDDDK (FLAG®) Chemiluminescent Western Blotting Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    RLKCA383
  • Supplier No.:
    KCA383
Spécifications
A propos de cet article
DYKDDDDK (FLAG®) Chemiluminescent Western Blotting Kit
  • This Chemiluminescent Western Blot Kit allows for the detection of FLAG® -tagged recombinant protein present in cell lysates provided by the user.
  • contains all of the necessary reagents
  • rapid proven protocol
  • extremely high signal detection of our luminol chemiluminescent substrate for the detection of FLAG® -tagged recombinant protein
The FLAG® epitope is a small but highly immunogenic peptide DYKDDDDK (N-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-C) which allows fusion proteins to retain their original conformation and function. The hydrophilic character of this sequence increases the likelihood that it will be located on the surface of FLAG® -tagged recombinant proteins for reaction with anti-DYKDDDDK (FLAG®) antibodies. The anti-DYKDDDDK (FLAG®) antibody included in this Western blot kit detects FLAG® -tagged recombinant proteins fused with FLAG® tag either at the amino-terminus or carboxy-terminus of the protein.
Use Rockland Immuno-chemicals’ Anti-DYKDDDDK (FLAG®) Chemiluminescent Kit for Western Blotting for detection of FLAG® -tagged recombinant proteins by Western blot. This kit is useful for both ''Western blotting'' and ''dot blotting'' methods. After protein separation and transfer, the membrane is probed with Anti-DYKDDDDK (FLAG®). Detection of the membrane bound antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems.
Customers Who Bought This Also Bought