ContactUsLinkComponent
Immunoprecipitation Kit: DYKDDDDK (FLAG®) FLAG® epitope tag, Rockland Immunochemicals
Immunoprecipitation Kit: DYKDDDDK (FLAG®) FLAG® epitope tag, Rockland Immunochemicals
# de catalogue RLKBA-319-383
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Immunoprecipitation Kit: DYKDDDDK (FLAG®) FLAG® epitope tag, Rockland Immunochemicals
# de catalogue RLKBA-319-383
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Immunoprecipitation Kit: DYKDDDDK (FLAG®) FLAG® epitope tag
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    RLKBA-319-383
  • Supplier No.:
    KBA-319-383
Spécifications
A propos de cet article
Rockland Immunochemicals' Anti-DYKDDDDK (FLAG®) Kit for Immunoprecipitation
  • rapid and efficient immunoprecipitation and elution of an active FLAG®-tagged recombinant protein in less than 2 hours
  • immunoprecipitation is performed with anti-FLAG® antibody coupled to agarose beads, which are generated by covalently linking agarose to a highly specific mouse monoclonal antibody raised against FLAG®
  • This kit is sufficient to perform 50 X 20 µL reactions and is stable for at least 1 year when stored as indicated.
Rockland Immunochemicals' Anti-DYKDDDDK (FLAG®) Kit for Immunoprecipitation allows for the purification by immunoprecipitation of recombinant proteins containing the FLAG® epitope tag provided by the user. The kit relies upon the high specificity of monoclonal antibody raised against the FLAG® epitope tag. This method is far easier and less costly than using antibodies produced against the recombinant protein itself therefore saving time and resources. Using the agarose bound antibody in this kit allows for efficient binding of FLAG® tag proteins without the need for preliminary steps or calibration. The immunoprecipitated FLAG® tag protein can be efficiently eluted from the agarose beads using a low pH elution step. The user is able to further characterize the resultant purified protein by size, post-translational modification, western blot and other assays.
Rockland Immunochemicals' Anti-DYKDDDDK (FLAG®) Kit for Immunoprecipitation is intended to provide a simple, reliable and convenient purification system for recombinant proteins containing the FLAG® epitope tag. Immunoprecipitation is a powerful technique for the isolation of proteins or protein complexes. Immunoprecipitation consists of several steps including cell lysis, binding of specific antigen to an antibody, antibody-antigen complex precipitation, precipitant wash steps and the dissociation of antigen from the complex. The FLAG® epitope tag is a small but highly immunogenic peptide DYKDDDDK (N-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-C), which allows fusion proteins to retain their original conformation and function. The hydrophilic character of FLAG® increases the likelihood that it will be located on the surface of the fusion protein where it is accessible to antibodies. Rockland Immunochemicals' Anti-DYKDDDDK (FLAG®) Kit for Immunoprecipitation allows a rapid and efficient immunoprecipitation and elution of an active FLAG® -tagged recombinant protein in less than 2 hours. The immunoprecipitation is performed with anti-FLAG® antibody coupled to agarose beads, which are generated by covalently linking agarose to a highly specific mouse monoclonal antibody raised against FLAG®. The provided protocol is a guideline. Any procedure can be altered according to specific experimental requirements.
Customers Who Bought This Also Bought