A propos de cet article
TrueBlot® Anti-Rabbit IgG Magnetic Beads
- used for separation and purification
- immunoassays, immunoprecipitation, and IP Western blots
- Bead mean diameter is ~0.5 μm, bead concentration is 5 mg/mL.
Trueblot® Magnetic Beads are uniform, non-aggregating, super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The super-paramagnetic nanoparticles are coupled with a biomolecule, such as goat anti-rabbit IgG, and are specifically designed, tested and quality controlled for isolation and purification of rabbit IgG, and immunoprecipitation methods using manual or automatic platforms. This antibody binds the heavy chain of all rabbit IgG subclasses and is suitable for immunoassays that utilize a rabbit IgG primary polyclonal antibody. Cell separation and sorting can be achieved using a rabbit IgG antibody to defined cell surface antigens. The beads have a large surface area with high capture efficiencies. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.
Trueblot® goat anti-rabbit IgG magnetic beads can be used for separation and purification of rabbit antibodies from serum or rabbit antibody-labeled components, as well as for immunoassays, immunoprecipitation, and IP Western blots. For antibody purification, goat anti-rabbit IgG magnetic beads are incubated with the rabbit antibody solution and then separated by magnets. After the unbound particulates are washed from the beads, the bound antibodies are eluted from the beads using the elution buffer. The beads are then magnetically separated from the eluted solution, which is removed manually. For IP, target specific antibody is incubated with goat anti-rabbit IgG magnetic beads. The unbound antibody is washed and the sample containing target antigen is added. After unbound particulates are washed from the beads, the purified protein is eluted from the beads using elution buffer. The samples are then resolved by SDS-PAGE and analyzed by Western blotting.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76221-844
- 76221-842
- Shipping conditions:
- Wet Ice
- Wet Ice