Paleozoic Fossil Collection
# de catalogue 470111-002
Fournisseur: SCOTT RESOURCES 2, LLC SE
Spécifications
- Description:Paleozoic fossil collection
- Cat. no.:470111-002
- Supplier No.:3110-S
Spécifications
A propos de cet article
During the Cambrian through the Permian periods, the fossil record shows the development of invertebrates, fish, land plants, insects, amphibians, and reptiles.
This 15-specimen set illustrates key life forms that dominated the Paleozoic era.
Delivery: The collection includes with Arthropod (1), Echinoderm (1), Corals (3), Mollusks (5), Brachiopods (2), Sponge (1), Plant (1) and Protozoan (1).
Customers Who Bought This Also Bought