Gilian® GilAir-3 Air Sampling Pumps, Sensidyne Inc
Fournisseur: Sensidyne
Certificats
A propos de cet article
Constant flow air sampling pump features clear external filter housing allows an immediate visual inspection of filter’s condition. The flow fault indicator lights up if unit operates outside ±5% of set flow rate. Battery test indicator lights up when battery can provide a minimum of 8 hours of operation over operating range.
- Constant low flow module allows sampling from 5 to 500 cc/min
- Constant pressure (multiflow) module allows sampling from 1 to 750 cc/min
Basic model is easy to use, clock model provides elapsed-time clock display and the program model provides auto start-stop and delayed start times up to 9999 minutes.
Delivery: Supplied with 36" tubing, air boss kit, reversible screwdriver and user manual.
Customers Who Bought This Also Bought