Mobile Folding Booth Seating, AmTab
Fournisseur: AmTab Manufacturing Corporation
Certificats
A propos de cet article
Mobile folding booth packages are used every day throughout the world in multi-purpose environments. High-quality state-of-the-art booths and tables are safe, durable, easy to operate, and perfect for heavy-use and heavy-turn settings providing years of trusted service.
- Affordable long-term value
- 100% made in the USA
- 15 year warranty
Custom high-speed gas cylinder stabilizer assist for long-lasting opening and closing functions and much more.
Certifications: UL and ANSI/BIFMA certified.
Spécifications
- Catalog No:
- 76178-604
- 76178-602
- Model:
- 76178-604
- 76178-602
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Dimensions:
- 20W×60L×40"H (folded)
- 20W×48L×40"H (folded)
- Environmental certification:
- MAS GREEN
- MAS GREEN